Assay Details
| Gene Symbol: LOC9272729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758717.2 | XP_015614203.1 | 28 - 29 | 4439 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9272729| Gene Symbol: LOC9272729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758717.2 | XP_015614203.1 | 28 - 29 | 4439 | 77 | |
Set Membership: |
|