Assay Details
| Gene Symbol: LOC9270331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761784.2 | XP_015617270.1 | 2 - 3 | 310 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9270331| Gene Symbol: LOC9270331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761784.2 | XP_015617270.1 | 2 - 3 | 310 | 100 | |
Set Membership: |
|