Assay Details
| Gene Symbol: LOC4349829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759896.2 | XP_015615382.1 | 1 - 2 | 1089 | 82 | |
| GenBank mRNA | AK071072.1 | - | 1 - 2 | 1067 | 82 | |
More Information
Set Membership: |
|

LOC4349829
Genomic Map