Assay Details
| Gene Symbol: LOC4351581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763045.2 | XP_015618531.1 | 1 - 2 | 194 | 59 | |
| GenBank mRNA | AK069765.1 | - | 1 - 2 | 133 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015763045.2 transcripts.
Set Membership: |
|

LOC4351581
Genomic Map