Assay Details
| Gene Symbol: LOC4351581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763045.2 | XP_015618531.1 | 7 - 8 | 1143 | 139 | |
| GenBank mRNA | AK069765.1 | - | 7 - 8 | 1082 | 139 | |
More Information
Set Membership: |
|

LOC4351581
Genomic Map