Assay Details
| Gene Symbol: LOC4351922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764056.1 | XP_015619542.1 | 1 - 2 | 165 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015764056.1 transcripts.
Set Membership: |
|

LOC4351922
Genomic Map