Assay Details
| Gene Symbol: LOC4352409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765065.2 | XP_015620551.1 | 1 - 2 | 529 | 103 | |
| GenBank mRNA | AK071593.1 | - | 1 - 2 | 471 | 103 | |
More Information
Set Membership: |
|

LOC4352409
Genomic Map