Assay Details
| Gene Symbol: LOC4352409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765065.2 | XP_015620551.1 | 3 - 4 | 824 | 76 | |
| GenBank mRNA | AK071593.1 | - | 3 - 4 | 766 | 76 | |
More Information
Set Membership: |
|

LOC4352409
Genomic Map