Assay Details
| Gene Symbol: LOC4352740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764789.1 | XP_015620275.1 | 6 - 7 | 1793 | 77 | |
| GenBank mRNA | AK073942.1 | - | 6 - 7 | 1800 | 77 | |
More Information
Set Membership: |
|

LOC4352740
Genomic Map