Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 10 - 11 | 1735 | 134 | |
| GenBank mRNA | AK065608.1 | - | 3 - 4 | 142 | 134 | |
| AY328087.1 | - | 10 - 11 | 1061 | 134 | ||
| AY785147.1 | - | 10 - 11 | 919 | 134 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map