Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 12 - 13 | 1975 | 81 | |
| GenBank mRNA | AK065608.1 | - | 4 - 5 | 664 | 81 | |
| AY328087.1 | - | 12 - 13 | 1301 | 81 | ||
| AY785147.1 | - | 12 - 13 | 1159 | 81 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map