Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 13 - 14 | 2022 | 77 | |
| GenBank mRNA | AK065608.1 | - | 5 - 6 | 711 | 77 | |
| AY328087.1 | - | 13 - 14 | 1348 | 77 | ||
| AY785147.1 | - | 13 - 14 | 1206 | 77 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map