Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 15 - 16 | 2375 | 59 | |
| GenBank mRNA | AK065608.1 | - | 7 - 8 | 1064 | 59 | |
| AY328087.1 | - | 15 - 16 | 1701 | 59 | ||
| AY785147.1 | - | 15 - 16 | 1559 | 59 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map