Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 19 - 20 | 3124 | 71 | |
| GenBank mRNA | AK065608.1 | - | 11 - 12 | 1813 | 71 | |
| AY785147.1 | - | 19 - 20 | 2308 | 71 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map