Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 1 - 2 | 971 | 86 | |
| GenBank mRNA | AY328087.1 | - | 1 - 2 | 297 | 86 | |
| AY785147.1 | - | 1 - 2 | 155 | 86 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map