Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 2 - 3 | 1023 | 78 | |
| GenBank mRNA | AY328087.1 | - | 2 - 3 | 349 | 78 | |
| AY785147.1 | - | 2 - 3 | 207 | 78 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map