Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 4 - 5 | 1197 | 101 | |
| GenBank mRNA | AY328087.1 | - | 4 - 5 | 523 | 101 | |
| AY785147.1 | - | 4 - 5 | 381 | 101 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map