Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 5 - 6 | 1349 | 143 | |
| GenBank mRNA | AY328087.1 | - | 5 - 6 | 675 | 143 | |
| AY785147.1 | - | 5 - 6 | 533 | 143 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map