Assay Details
| Gene Symbol: LOC4352928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763865.2 | XP_015619351.1 | 9 - 10 | 1691 | 95 | |
| GenBank mRNA | AK065608.1 | - | 2 - 3 | 98 | 95 | |
| AY328087.1 | - | 9 - 10 | 1017 | 95 | ||
| AY785147.1 | - | 9 - 10 | 875 | 95 | ||
More Information
Set Membership: |
|

LOC4352928
Genomic Map