Assay Details
| Gene Symbol: LOC4350403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761965.2 | XP_015617451.1 | 7 - 8 | 1755 | 71 | |
| GenBank mRNA | AK067849.1 | - | 3 - 4 | 298 | 71 | |
More Information
Set Membership: |
|

LOC4350403
Genomic Map