Assay Details
| Gene Symbol: LOC4350403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761965.2 | XP_015617451.1 | 10 - 11 | 2263 | 67 | |
| GenBank mRNA | AK067849.1 | - | 6 - 7 | 806 | 67 | |
More Information
Set Membership: |
|

LOC4350403
Genomic Map