Assay Details
| Gene Symbol: LOC4350685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759653.2 | XP_015615139.1 | 1 - 2 | 153 | 80 | |
| GenBank mRNA | AK072844.1 | - | 1 - 2 | 85 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015759653.2 transcripts.
Set Membership: |
|

LOC4350685
Genomic Map