Assay Details
| Gene Symbol: LOC4350685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759653.2 | XP_015615139.1 | 3 - 4 | 310 | 91 | |
| GenBank mRNA | AK072844.1 | - | 3 - 4 | 242 | 91 | |
| HQ858816.1 | - | 3 - 4 | 196 | 91 | ||
More Information
Set Membership: |
|

LOC4350685
Genomic Map