Assay Details
| Gene Symbol: LOC4350712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760120.2 | XP_015615606.1 | 1 - 2 | 439 | 158 | |
| GenBank mRNA | AK287462.1 | - | 1 - 2 | 1103 | 158 | |
More Information
Set Membership: |
|

LOC4350712
Genomic Map