Assay Details
| Gene Symbol: LOC4351224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761411.1 | XP_015616897.1 | 2 - 3 | 460 | 72 | |
| GenBank mRNA | AB034698.1 | - | 2 - 3 | 396 | 72 | |
| AB110180.1 | - | 2 - 3 | 361 | 72 | ||
| AK060841.1 | - | 1 - 2 | 181 | 72 | ||
| AK060847.1 | - | 2 - 3 | 397 | 72 | ||
| AK064986.1 | - | 2 - 3 | 398 | 72 | ||
| AK119513.1 | - | 2 - 3 | 409 | 72 | ||
| HQ324826.1 | - | 2 - 3 | 377 | 72 | ||
| KC140121.1 | - | 2 - 3 | 369 | 72 | ||
| U74321.1 | - | 2 - 3 | 369 | 72 | ||
More Information
Set Membership: |
|

LOC4351224
Genomic Map