Assay Details
| Gene Symbol: LOC4326159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792146.2 | XP_015647632.1 | 6 - 7 | 1440 | 93 | |
| GenBank mRNA | AK058708.1 | - | 1 - 2 | 243 | 93 | |
| AK070363.1 | - | 6 - 7 | 1443 | 93 | ||
| AK287579.1 | - | 3542 | 93 | |||
More Information
Set Membership: |
|

LOC4326159
Genomic Map