Assay Details
| Gene Symbol: LOC4326159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792146.2 | XP_015647632.1 | 8 - 9 | 1703 | 113 | |
| GenBank mRNA | AK058708.1 | - | 3 - 4 | 506 | 113 | |
| AK070363.1 | - | 8 - 9 | 1706 | 113 | ||
| AK287579.1 | - | 3805 | 113 | |||
More Information
Set Membership: |
|

LOC4326159
Genomic Map