Assay Details
| Gene Symbol: LOC4326159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792146.2 | XP_015647632.1 | 10 - 11 | 1875 | 68 | |
| GenBank mRNA | AK058708.1 | - | 5 - 6 | 678 | 68 | |
| AK070363.1 | - | 10 - 11 | 1878 | 68 | ||
| AK287579.1 | - | 3977 | 68 | |||
More Information
Set Membership: |
|

LOC4326159
Genomic Map