Assay Details
| Gene Symbol: LOC9271572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777004.2 | XP_015632490.1 | 5 - 6 | 770 | 82 | |
| XM_015777012.2 | XP_015632498.1 | 5 - 6 | 767 | 82 | ||
| GenBank mRNA | AK108568.1 | - | 5 - 6 | 805 | 82 | |
| AY553724.1 | - | 4 - 5 | 549 | 82 | ||
More Information
Set Membership: |
|

LOC9271572
Genomic Map