Assay Details
| Gene Symbol: LOC9271572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777004.2 | XP_015632490.1 | 7 - 8 | 851 | 62 | |
| XM_015777012.2 | XP_015632498.1 | 7 - 8 | 848 | 62 | ||
| GenBank mRNA | AK108568.1 | - | 7 - 8 | 886 | 62 | |
| AY553724.1 | - | 6 - 7 | 630 | 62 | ||
More Information
Set Membership: |
|

LOC9271572
Genomic Map