Assay Details
| Gene Symbol: LOC9271572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777004.2 | XP_015632490.1 | 9 - 10 | 1090 | 85 | |
| XM_015777012.2 | XP_015632498.1 | 9 - 10 | 1087 | 85 | ||
| GenBank mRNA | AK108568.1 | - | 9 - 10 | 1125 | 85 | |
More Information
Set Membership: |
|

LOC9271572
Genomic Map