Assay Details
| Gene Symbol: LOC4327786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778050.2 | XP_015633536.1 | 6 - 7 | 1135 | 75 | |
| GenBank mRNA | AK064408.1 | - | 6 - 7 | 970 | 75 | |
More Information
Set Membership: |
|

LOC4327786
Genomic Map