Assay Details
| Gene Symbol: LOC4326901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766623.2 | XP_015622109.1 | 1 - 2 | 564 | 87 | |
| GenBank mRNA | AK058287.1 | - | 1 - 2 | 507 | 87 | |
| AK059435.1 | - | 1 - 2 | 511 | 87 | ||
| AK069333.1 | - | 1 - 2 | 521 | 87 | ||
| FJ940751.1 | - | 1 - 2 | 473 | 87 | ||
More Information
Set Membership: |
|

LOC4326901
Genomic Map