Assay Details
| Gene Symbol: LOC4326901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766623.2 | XP_015622109.1 | 2 - 3 | 631 | 85 | |
| GenBank mRNA | AK058287.1 | - | 2 - 3 | 574 | 85 | |
| AK059435.1 | - | 2 - 3 | 578 | 85 | ||
| AK069333.1 | - | 2 - 3 | 588 | 85 | ||
| FJ940751.1 | - | 2 - 3 | 540 | 85 | ||
More Information
Set Membership: |
|

LOC4326901
Genomic Map