Assay Details
| Gene Symbol: LOC4326901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766623.2 | XP_015622109.1 | 3 - 4 | 956 | 95 | |
| GenBank mRNA | AK058287.1 | - | 3 - 4 | 899 | 95 | |
| AK059435.1 | - | 3 - 4 | 903 | 95 | ||
| AK069333.1 | - | 3 - 4 | 913 | 95 | ||
| FJ940751.1 | - | 3 - 4 | 865 | 95 | ||
More Information
Set Membership: |
|

LOC4326901
Genomic Map