Assay Details
| Gene Symbol: LOC4327772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766254.1 | XP_015621740.1 | 10 - 11 | 2753 | 77 | |
| GenBank mRNA | AK062171.1 | - | 4 - 5 | 610 | 77 | |
| AK103288.1 | - | 10 - 11 | 2756 | 77 | ||
More Information
Set Membership: |
|

LOC4327772
Genomic Map