Assay Details
| Gene Symbol: LOC4326479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774536.2 | XP_015630022.1 | 4 - 5 | 788 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4326479| Gene Symbol: LOC4326479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774536.2 | XP_015630022.1 | 4 - 5 | 788 | 91 | |
Set Membership: |
|