Assay Details
| Gene Symbol: LOC4326288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766376.2 | XP_015621862.1 | 1 - 2 | 136 | 99 | |
| GenBank mRNA | AK070843.1 | - | 1 - 2 | 143 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766376.2 transcripts.
Set Membership: |
|

LOC4326288
Genomic Map