Assay Details
| Gene Symbol: LOC4326288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766376.2 | XP_015621862.1 | 3 - 4 | 421 | 147 | |
| XM_015766379.2 | XP_015621865.1 | 2 - 3 | 285 | 147 | ||
| GenBank mRNA | AK070843.1 | - | 3 - 4 | 428 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766379.2 transcripts.
Set Membership: |
|

LOC4326288
Genomic Map