Assay Details
| Gene Symbol: LOC4326288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766376.2 | XP_015621862.1 | 4 - 5 | 498 | 150 | |
| XM_015766379.2 | XP_015621865.1 | 3 - 4 | 362 | 150 | ||
| GenBank mRNA | AK070843.1 | - | 4 - 5 | 505 | 150 | |
More Information
Set Membership: |
|

LOC4326288
Genomic Map