Assay Details
| Gene Symbol: LOC4326288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766376.2 | XP_015621862.1 | 5 - 6 | 606 | 80 | |
| XM_015766379.2 | XP_015621865.1 | 4 - 5 | 470 | 80 | ||
| GenBank mRNA | AK070843.1 | - | 5 - 6 | 613 | 80 | |
More Information
Set Membership: |
|

LOC4326288
Genomic Map