Assay Details
| Gene Symbol: LOC4327935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765875.1 | XP_015621361.1 | 1 - 2 | 162 | 73 | |
| GenBank mRNA | AK061158.1 | - | 1 - 2 | 127 | 73 | |
| AK073996.1 | - | 1 - 2 | 130 | 73 | ||
More Information
Set Membership: |
|

LOC4327935
Genomic Map