Assay Details
| Gene Symbol: LOC4327935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765875.1 | XP_015621361.1 | 4 - 5 | 571 | 89 | |
| GenBank mRNA | AK061158.1 | - | 4 - 5 | 536 | 89 | |
| AK073996.1 | - | 4 - 5 | 539 | 89 | ||
More Information
Set Membership: |
|

LOC4327935
Genomic Map