Assay Details
| Gene Symbol: LOC4327687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756487.2 | XP_015611973.1 | 1 - 2 | 529 | 91 | |
| GenBank mRNA | AK066220.1 | - | 1 - 2 | 458 | 91 | |
More Information
Set Membership: |
|

LOC4327687
Genomic Map