Assay Details
| Gene Symbol: LOC4327687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756487.2 | XP_015611973.1 | 4 - 5 | 964 | 96 | |
| GenBank mRNA | AK066220.1 | - | 4 - 5 | 893 | 96 | |
More Information
Set Membership: |
|

LOC4327687
Genomic Map