Assay Details
| Gene Symbol: LOC4327687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756487.2 | XP_015611973.1 | 6 - 7 | 1272 | 80 | |
| GenBank mRNA | AK066220.1 | - | 6 - 7 | 1201 | 80 | |
More Information
Set Membership: |
|

LOC4327687
Genomic Map