Assay Details
| Gene Symbol: LOC4327564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793412.2 | XP_015648898.1 | 1 - 2 | 435 | 68 | |
| GenBank mRNA | AF466358.1 | - | 1 - 2 | 306 | 68 | |
| AK070798.1 | - | 1 - 2 | 345 | 68 | ||
| AK099315.1 | - | 1 - 2 | 343 | 68 | ||
More Information
Set Membership: |
|

LOC4327564
Genomic Map