Assay Details
| Gene Symbol: LOC4327564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793412.2 | XP_015648898.1 | 2 - 3 | 507 | 144 | |
| GenBank mRNA | AF466358.1 | - | 2 - 3 | 378 | 144 | |
| AK070798.1 | - | 2 - 3 | 417 | 144 | ||
| AK099315.1 | - | 2 - 3 | 415 | 144 | ||
More Information
Set Membership: |
|

LOC4327564
Genomic Map