Assay Details
| Gene Symbol: LOC4327564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793412.2 | XP_015648898.1 | 3 - 4 | 654 | 78 | |
| GenBank mRNA | AF466358.1 | - | 3 - 4 | 525 | 78 | |
| AK070798.1 | - | 3 - 4 | 564 | 78 | ||
| AK099315.1 | - | 3 - 4 | 562 | 78 | ||
More Information
Set Membership: |
|

LOC4327564
Genomic Map