Assay Details
| Gene Symbol: LOC4327431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763095.2 | 1 | 1158 | 127 | ||
| GenBank mRNA | AB115668.1 | - | 1 | 848 | 127 | |
| JN210896.1 | - | 1 | 1164 | 127 | ||
More Information
Set Membership: |
|

LOC4327431
Genomic Map